Assay Design
Gene Transcripts
- Gene Symbol
- set-14
- Entrez Gene ID
-
174662
- Gene Name
- SET domain-containing protein 14
- Gene Aliases
-
CELE_R06F6.4
- Chromosome Location
-
Chr.II: 10793575 - 10796709 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063922.6 |
NP_496323.1 |
2 - 3 |
447 |
83 |
View Full Product Details