Assay Design
Gene Transcripts
- Gene Symbol
- sra-11
- Entrez Gene ID
-
191782
- Gene Name
- Serpentine receptor class alpha-11
- Gene Aliases
-
CELE_F44F4.13
- Chromosome Location
-
Chr.II: 10908808 - 10910319 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063947.2 |
NP_496348.2 |
2 - 3 |
461 |
81 |
View Full Product Details