Assay Design
Gene Transcripts
- Gene Symbol
- ebp-2
- Entrez Gene ID
-
174744
- Gene Name
- microtubule End Binding Protein
- Gene Aliases
-
CELE_VW02B12L.3
- Chromosome Location
-
Chr.II: 11442725 - 11444016 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064037.6 |
NP_496438.1 |
2 - 3 |
346 |
70 |
View Full Product Details