Assay Design
Gene Transcripts
- Gene Symbol
- adbp-1
- Entrez Gene ID
-
174745
- Gene Name
- Adr-2-binding protein 1
- Gene Aliases
-
CELE_VW02B12L.4
- Chromosome Location
-
Chr.II: 11444162 - 11445255 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064038.8 |
NP_496439.1 |
5 - 6 |
590 |
68 |
GenBank mRNA |
AF303256.1 |
- |
5 - 6 |
590 |
68 |
View Full Product Details