Assay Design



Gene Transcripts

Gene Symbol
twk-5
Entrez Gene ID
174756
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_B0334.2
Chromosome Location
Chr.II: 11481080 - 11485300 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026725.6 NP_001021896.1 3 - 4 562 65

View Full Product Details