Assay Design
Gene Transcripts
- Gene Symbol
- npp-5
- Entrez Gene ID
-
174779
- Gene Name
- Nuclear pore complex protein 5
- Gene Aliases
-
CELE_F07A11.3
- Chromosome Location
-
Chr.II: 11596767 - 11599809 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064080.6 |
NP_496481.1 |
2 - 3 |
218 |
77 |
View Full Product Details