Assay Design
Gene Transcripts
- Gene Symbol
- sra-14
- Entrez Gene ID
-
185262
- Gene Name
- Serpentine Receptor, class A (alpha)
- Gene Aliases
-
CELE_F35C5.2
- Chromosome Location
-
Chr.II: 12885272 - 12888608 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064337.7 |
NP_496738.2 |
2 - 3 |
386 |
89 |
View Full Product Details