Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083166.5 | NP_001076635.1 | 10 - 11 | 1765 | 111 |
NM_001083167.2 | NP_001076636.1 | 6 - 7 | 1285 | 111 | |
NM_001129183.4 | NP_001122655.1 | 11 - 12 | 1915 | 111 | |
NM_001267434.3 | NP_001254363.1 | 4 - 5 | 718 | 111 | |
NM_001313556.2 | NP_001300485.1 | 11 - 12 | 1918 | 111 | |
NM_001313557.3 | NP_001300486.1 | 10 - 10 | 1714 | 111 | |
GenBank mRNA | DQ523832.1 | - | 10 - 11 | 1765 | 111 |