Assay Design



Gene Transcripts

Gene Symbol
alh-7
Entrez Gene ID
174991
Gene Name
protein-coding
Gene Aliases
CELE_F45H10.1
Chromosome Location
Chr.II: 13485758 - 13491786 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267464.2 NP_001254393.1 3 - 4 1068 101
NM_001381642.1 1 - 2 192 101

View Full Product Details