Assay Design
Gene Transcripts
- Gene Symbol
- sre-43
- Entrez Gene ID
-
189282
- Gene Name
- Serpentine Receptor, class E (epsilon)
- Gene Aliases
-
CELE_W07G1.6
- Chromosome Location
-
Chr.II: 13966534 - 13969079 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064522.1 |
NP_496923.1 |
3 - 4 |
626 |
120 |
View Full Product Details