Assay Design
Gene Transcripts
- Gene Symbol
- daf-5
- Entrez Gene ID
-
175054
- Gene Name
- c-SKI_SMAD_bind domain-containing protein
- Gene Aliases
-
CELE_W01G7.1
- Chromosome Location
-
Chr.II: 14033216 - 14037721 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064540.6 |
NP_496941.1 |
3 - 4 |
557 |
82 |
GenBank mRNA |
AY438643.1 |
- |
3 - 4 |
557 |
82 |
View Full Product Details