Assay Design
Gene Transcripts
- Gene Symbol
- imb-2
- Entrez Gene ID
-
175094
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_R06A4.4
- Chromosome Location
-
Chr.II: 14365701 - 14373901 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064587.5 |
NP_496988.1 |
1 - 2 |
73 |
80 |
View Full Product Details