Assay Design



Gene Transcripts

Gene Symbol
gcy-29
Entrez Gene ID
175116
Gene Name
Receptor-type guanylate cyclase gcy-29
Gene Aliases
CELE_C04H5.3
Chromosome Location
Chr.II: 14540895 - 14548791 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377902.1 NP_001364597.1 4 - 5 687 99

View Full Product Details