Assay Design



Gene Transcripts

Gene Symbol
cog-1
Entrez Gene ID
175149
Gene Name
protein-coding
Gene Aliases
CELE_R03C1.3
Chromosome Location
Chr.II: 14902491 - 14907535 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027093.3 NP_001022264.1 1 - 2 334 68
GenBank mRNA AY151209.1 - 1 - 2 334 68

View Full Product Details