Assay Design
Gene Transcripts
- Gene Symbol
- spt-4
- Entrez Gene ID
-
175172
- Gene Name
- Transcription elongation factor SPT4
- Gene Aliases
-
CELE_F54C4.2
- Chromosome Location
-
Chr.III: 69073 - 69752 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064734.6 |
NP_497135.1 |
1 - 2 |
175 |
59 |
View Full Product Details