Assay Design
Gene Transcripts
- Gene Symbol
- klp-20
- Entrez Gene ID
-
3565057
- Gene Name
- Kinesin-like protein klp-20
- Gene Aliases
-
CELE_Y50D7A.6
- Chromosome Location
-
Chr.III: 194460 - 228838 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064777.6 |
NP_497178.1 |
11 - 12 |
1534 |
72 |
- Gene Symbol
- F40G9.17
- Entrez Gene ID
-
13188172
- Gene Name
- ANK_REP_REGION domain-containing protein
- Gene Aliases
-
CELE_F40G9.17
- Chromosome Location
-
Chr.III: 194460 - 235089 on Build WBcel235
- Gene Symbol
- hpo-38
- Entrez Gene ID
-
175188
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_Y50D7A.5
- Chromosome Location
-
Chr.III: 194460 - 235092 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001384002.1 |
|
13 - 14 |
1894 |
72 |
View Full Product Details