Assay Design



Gene Transcripts

Gene Symbol
ptl-1
Entrez Gene ID
175230
Gene Name
protein-coding
Gene Aliases
CELE_F42G9.9
Chromosome Location
Chr.III: 763810 - 771391 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381708.2 NP_001367354.1 6 - 7 1205 153
GenBank mRNA U67967.1 - 6 - 7 1203 153

View Full Product Details