Assay Design
Gene Transcripts
- Gene Symbol
- tag-51
- Entrez Gene ID
-
175245
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_K02F3.1
- Chromosome Location
-
Chr.III: 854221 - 860072 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064876.9 |
NP_497277.2 |
2 - 3 |
462 |
71 |
View Full Product Details