Assay Design



Gene Transcripts

Gene Symbol
trp-2
Entrez Gene ID
192056
Gene Name
protein-coding
Gene Aliases
CELE_R06B10.4
Chromosome Location
Chr.III: 983261 - 1001931 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027532.5 NP_001022703.1 5 - 6 916 70
NM_064910.8 NP_497311.2 11 - 12 2175 70
GenBank mRNA AJ276028.1 - 10 - 11 2005 70

View Full Product Details