Assay Design



Gene Transcripts

Gene Symbol
trp-2
Entrez Gene ID
192056
Gene Name
protein-coding
Gene Aliases
CELE_R06B10.4
Chromosome Location
Chr.III: 983261 - 1001931 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027532.5 NP_001022703.1 2 - 3 284 98
NM_064910.8 NP_497311.2 8 - 9 1543 98
GenBank mRNA AJ276028.1 - 7 - 8 1373 98

View Full Product Details