Assay Design



Gene Transcripts

Gene Symbol
uaf-1
Entrez Gene ID
175270
Gene Name
protein-coding
Gene Aliases
CELE_Y92C3B.2
Chromosome Location
Chr.III: 1062794 - 1076555 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027796.4 NP_001022967.1 1 - 2 232 64
GenBank mRNA U79157.1 - 1 - 2 232 64

View Full Product Details