Assay Design
Gene Transcripts
- Gene Symbol
- fbxa-28
- Entrez Gene ID
-
191033
- Gene Name
- F-box domain-containing protein
- Gene Aliases
-
CELE_Y119D3A.2
- Chromosome Location
-
Chr.III: 1297904 - 1299484 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064975.1 |
NP_497376.1 |
1 - 2 |
55 |
86 |
View Full Product Details