Assay Design
Gene Transcripts
- Gene Symbol
- fbxa-75
- Entrez Gene ID
-
191032
- Gene Name
- FTH domain-containing protein
- Gene Aliases
-
CELE_Y119D3A.1
- Chromosome Location
-
Chr.III: 1303714 - 1305461 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064976.1 |
NP_497377.1 |
2 - 3 |
711 |
101 |
View Full Product Details