Assay Design
Gene Transcripts
- Gene Symbol
- fbxa-14
- Entrez Gene ID
-
175294
- Gene Name
- F-box domain-containing protein
- Gene Aliases
-
CELE_Y82E9BL.10
- Chromosome Location
-
Chr.III: 1322124 - 1324633 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064983.6 |
NP_497384.1 |
3 - 4 |
1211 |
123 |
View Full Product Details