Assay Design
Gene Transcripts
- Gene Symbol
- glb-30
- Entrez Gene ID
-
189513
- Gene Name
- GLOBIN domain-containing protein
- Gene Aliases
-
CELE_Y22D7AR.5
- Chromosome Location
-
Chr.III: 1665831 - 1668634 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065041.4 |
NP_497442.2 |
6 - 7 |
552 |
73 |
GenBank mRNA |
EF471979.1 |
- |
6 - 7 |
537 |
73 |
View Full Product Details