Assay Design
Gene Transcripts
- Gene Symbol
- frl-1
- Entrez Gene ID
-
175346
- Gene Name
- FRL (Formin Related gene in Leukocytes) homolog
- Gene Aliases
-
CELE_Y48G9A.4
- Chromosome Location
-
Chr.III: 2124137 - 2151554 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065104.7 |
NP_497505.3 |
3 - 4 |
337 |
77 |
View Full Product Details