Assay Design



Gene Transcripts

Gene Symbol
ceh-44
Entrez Gene ID
175372
Gene Name
protein-coding
Gene Aliases
CELE_Y54F10AM.4
Chromosome Location
Chr.III: 2530606 - 2546420 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065174.7 NP_497575.2 6 - 7 1070 88
NM_065175.7 NP_497576.1 6 - 7 1061 88
NM_065176.6 NP_497577.2 6 - 7 1061 88

View Full Product Details