Assay Design



Gene Transcripts

Gene Symbol
sft-1
Entrez Gene ID
175415
Gene Name
protein-coding
Gene Aliases
CELE_H06I04.2
Chromosome Location
Chr.III: 3086671 - 3091737 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001312947.3 NP_001299876.1 3 - 4 574 102
NM_065256.6 NP_497657.1 4 - 5 866 102

View Full Product Details