Assay Design



Gene Transcripts

Gene Symbol
ral-1
Entrez Gene ID
175433
Gene Name
protein-coding
Gene Aliases
CELE_Y53G8AR.3
Chromosome Location
Chr.III: 3311310 - 3317716 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267937.3 NP_001254866.1 3 - 4 533 67
NM_001267938.2 NP_001254867.1 2 - 3 125 67

View Full Product Details