Assay Design
Gene Transcripts
- Gene Symbol
- ptp-1
- Entrez Gene ID
-
175463
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C48D5.2
- Chromosome Location
-
Chr.III: 3590722 - 3611971 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065331.4 |
NP_497732.1 |
5 - 6 |
892 |
111 |
View Full Product Details