Assay Design
Gene Transcripts
- Gene Symbol
- lin-48
- Entrez Gene ID
-
175485
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_F34D10.5
- Chromosome Location
-
Chr.III: 3745438 - 3747572 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065358.5 |
NP_497759.1 |
4 - 5 |
576 |
86 |
GenBank mRNA |
AF134806.1 |
- |
4 - 5 |
578 |
86 |
View Full Product Details