Assay Design



Gene Transcripts

Gene Symbol
mtm-3
Entrez Gene ID
175490
Gene Name
protein-coding
Gene Aliases
CELE_T24A11.1
Chromosome Location
Chr.III: 3789982 - 3800547 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027623.6 NP_001022794.2 4 - 5 563 159
NM_065365.9 NP_497766.3 4 - 5 551 159
GenBank mRNA DQ988041.1 - 4 - 5 412 159

View Full Product Details