Assay Design
Gene Transcripts
- Gene Symbol
- dpy-27
- Entrez Gene ID
-
175492
- Gene Name
- Chromosome condensation protein dpy-27
- Gene Aliases
-
CELE_R13G10.1
- Chromosome Location
-
Chr.III: 3813018 - 3820991 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065370.6 |
NP_497771.1 |
11 - 12 |
3684 |
66 |
View Full Product Details