Assay Design
Gene Transcripts
- Gene Symbol
- amx-1
- Entrez Gene ID
-
175493
- Gene Name
- Amine oxidase family member 1
- Gene Aliases
-
CELE_R13G10.2
- Chromosome Location
-
Chr.III: 3822403 - 3826407 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065371.6 |
NP_497772.2 |
6 - 7 |
1474 |
92 |
View Full Product Details