Assay Design



Gene Transcripts

Gene Symbol
nth-1
Entrez Gene ID
187770
Gene Name
protein-coding
Gene Aliases
CELE_R10E4.5
Chromosome Location
Chr.III: 4291543 - 4294253 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267978.2 NP_001254907.1 1 - 2 178 68
NM_001384068.1 1 - 2 1032 68
GenBank mRNA AB518695.1 - 1 - 2 192 68

View Full Product Details