Assay Design



Gene Transcripts

Gene Symbol
nth-1
Entrez Gene ID
187770
Gene Name
protein-coding
Gene Aliases
CELE_R10E4.5
Chromosome Location
Chr.III: 4291543 - 4294253 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267978.2 NP_001254907.1 3 - 4 353 103
NM_001267979.3 NP_001254908.1 2 - 3 145 103
NM_001384068.1 3 - 4 1207 103
GenBank mRNA AB518695.1 - 3 - 4 367 103

View Full Product Details