Assay Design
Gene Transcripts
- Gene Symbol
- ceh-43
- Entrez Gene ID
-
175581
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C28A5.4
- Chromosome Location
-
Chr.III: 4446629 - 4449943 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065503.4 |
NP_497904.1 |
3 - 4 |
441 |
65 |
View Full Product Details