Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001027391.8 | NP_001022562.1 | 8 - 9 | 1299 | 76 |
NM_001027392.6 | NP_001022563.1 | 8 - 9 | 1299 | 76 | |
GenBank mRNA | AF256466.1 | - | 8 - 9 | 1289 | 76 |
AF256467.1 | - | 8 - 9 | 1289 | 76 |