Assay Design



Gene Transcripts

Gene Symbol
sel-2
Entrez Gene ID
175606
Gene Name
protein-coding
Gene Aliases
CELE_F10F2.1
Chromosome Location
Chr.III: 4596059 - 4612384 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377920.3 NP_001364559.1 21 - 22 5154 78
NM_065538.6 NP_497939.2 20 - 21 5019 78

View Full Product Details