Assay Design
Gene Transcripts
- Gene Symbol
- ppat-1
- Entrez Gene ID
-
175613
- Gene Name
- Amidophosphoribosyltransferase
- Gene Aliases
-
CELE_T04A8.5
- Chromosome Location
-
Chr.III: 4689280 - 4692171 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065557.8 |
NP_497958.1 |
2 - 3 |
565 |
126 |
View Full Product Details