Assay Design
Gene Transcripts
- Gene Symbol
- ras-2
- Entrez Gene ID
-
175625
- Gene Name
- R-RAS related
- Gene Aliases
-
CELE_F17C8.4
- Chromosome Location
-
Chr.III: 4737586 - 4740075 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065571.6 |
NP_497972.1 |
3 - 4 |
575 |
121 |
GenBank mRNA |
U80675.1 |
- |
3 - 4 |
570 |
121 |
View Full Product Details