Assay Design
Gene Transcripts
- Gene Symbol
- rbg-3
- Entrez Gene ID
-
175629
- Gene Name
- Rab-GAP TBC domain-containing protein
- Gene Aliases
-
CELE_B0393.2
- Chromosome Location
-
Chr.III: 4754424 - 4757021 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065578.6 |
NP_497979.1 |
5 - 6 |
1022 |
88 |
View Full Product Details