Assay Design



Gene Transcripts

Gene Symbol
pal-1
Entrez Gene ID
175638
Gene Name
protein-coding
Gene Aliases
CELE_C38D4.6
Chromosome Location
Chr.III: 4804688 - 4809622 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026038.6 NP_001021209.1 6 - 7 904 79
NM_001026039.6 NP_001021210.1 6 - 7 948 79
NM_001267992.4 NP_001254921.1 4 - 5 624 79

View Full Product Details