Assay Design
Gene Transcripts
- Gene Symbol
- rpb-2
- Entrez Gene ID
-
175668
- Gene Name
- DNA-directed RNA polymerase II subunit RPB2
- Gene Aliases
-
CELE_C26E6.4
- Chromosome Location
-
Chr.III: 4939567 - 4944404 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065646.8 |
NP_498047.2 |
2 - 3 |
1300 |
85 |
View Full Product Details