Assay Design



Gene Transcripts

Gene Symbol
cah-1
Entrez Gene ID
186231
Gene Name
protein-coding
Gene Aliases
CELE_F54D8.4
Chromosome Location
Chr.III: 5083206 - 5085629 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322586.2 NP_001309492.1 4 - 4 568 149

View Full Product Details