Assay Design
Gene Transcripts
- Gene Symbol
- tbx-2
- Entrez Gene ID
-
175698
- Gene Name
- T-box protein 2
- Gene Aliases
-
CELE_F21H11.3
- Chromosome Location
-
Chr.III: 5130923 - 5136319 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001382860.1 |
|
1 - 2 |
399 |
89 |
View Full Product Details