Assay Design
Gene Transcripts
- Gene Symbol
- atp-2
- Entrez Gene ID
-
175716
- Gene Name
- ATP synthase subunit beta, mitochondrial
- Gene Aliases
-
CELE_C34E10.6
- Chromosome Location
-
Chr.III: 5228417 - 5230544 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065710.7 |
NP_498111.2 |
4 - 5 |
1527 |
76 |
View Full Product Details