Assay Design
Gene Transcripts
- Gene Symbol
- nspb-12
- Entrez Gene ID
-
184261
- Gene Name
- Nematode Specific Peptide family, group B
- Gene Aliases
-
CELE_F09F7.8
- Chromosome Location
-
Chr.III: 5568213 - 5568842 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065793.5 |
NP_498194.2 |
2 - 2 |
181 |
70 |
View Full Product Details