Assay Design



Gene Transcripts

Gene Symbol
par-3
Entrez Gene ID
175783
Gene Name
protein-coding
Gene Aliases
CELE_F54E7.3
Chromosome Location
Chr.III: 5664229 - 5676795 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027436.3 NP_001022607.1 10 - 11 3132 125
NM_001306735.2 NP_001293664.1 6 - 7 2814 125
NM_001306740.3 NP_001293669.1 10 - 11 3132 125
NM_001379719.1 NP_001368259.1 8 - 9 2904 125
NM_001379721.1 NP_001368258.1 6 - 7 2814 125
NM_001379722.1 NP_001368260.1 5 - 6 2667 125
GenBank mRNA U25032.1 - 10 - 11 3131 125

View Full Product Details