Assay Design



Gene Transcripts

Gene Symbol
par-3
Entrez Gene ID
175783
Gene Name
protein-coding
Gene Aliases
CELE_F54E7.3
Chromosome Location
Chr.III: 5664229 - 5676795 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027436.3 NP_001022607.1 12 - 13 3562 83
NM_001306735.2 NP_001293664.1 9 - 10 3417 83
NM_001306736.3 NP_001293665.1 9 - 10 3408 83
NM_001306739.3 NP_001293668.1 13 - 14 3726 83
NM_001306740.3 NP_001293669.1 13 - 14 3735 83
NM_001306741.3 NP_001293670.1 11 - 12 3406 83
NM_001372593.2 NP_001360234.1 8 - 9 3235 83
NM_001372595.2 NP_001360237.1 7 - 8 3088 83
NM_001379719.1 NP_001368259.1 10 - 11 3334 83
NM_001379720.2 NP_001367341.1 8 - 9 3364 83
NM_001379721.1 NP_001368258.1 8 - 9 3244 83
NM_001379722.1 NP_001368260.1 7 - 8 3097 83
NM_001382875.1 12 - 13 3549 83
GenBank mRNA U25032.1 - 12 - 13 3561 83

View Full Product Details